ENA300792249 Enamine

3-[(2-aminophenyl)formamido]propanamide

  • MDL number MFCD12136331
  • Cas Number : 1156868-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 197...199
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67492

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67492
Availability details for
Quantity
  • Availability
  • Add to Cart