ENA300792252 Enamine

2-(1,1-dioxo-1lambda6-thiomorpholin-4-yl)-2-phenylacetic acid

  • MDL number MFCD02081980
  • Cas Number : 446276-14-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-67496

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67496
Availability details for
Quantity
  • Availability
  • Add to Cart