ENA300792253 Enamine

3-[(pyridin-3-yl)methoxy]pyridin-2-amine

  • MDL number MFCD10690776
  • Cas Number : 96428-82-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67498

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67498
Availability details for
Quantity
  • Availability
  • Add to Cart