ENA300792254 Enamine

2-(2,5-difluorophenyl)-2-hydroxyacetonitrile

  • MDL number MFCD12169396
  • Cas Number : 1155082-23-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67499

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67499
Availability details for
Quantity
  • Availability
  • Add to Cart