ENA300792256 Enamine

1-(methylamino)-2,3-dihydro-1H-indene-1-carboxylic acid

  • MDL number MFCD12047819
  • Cas Number : 1157935-94-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67501

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67501
Availability details for
Quantity
  • Availability
  • Add to Cart