ENA300792264 Enamine

3-amino-N-tert-butylpropanamide hydrochloride

  • MDL number MFCD11109909
  • Cas Number : 1103861-54-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 52...54
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67518

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67518
Availability details for
Quantity
  • Availability
  • Add to Cart