ENA300792265 Enamine

3-fluoro-4-(morpholin-4-yl)aniline

  • MDL number MFCD02571270
  • Cas Number : 93246-53-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121...123
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67519

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67519
Availability details for
Quantity
  • Availability
  • Add to Cart