ENA300792266 Enamine

2-chloroimidazo[1,2-a]pyridine-3-carbonitrile

  • MDL number MFCD09041343
  • Cas Number : 131773-48-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67520

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67520
Availability details for
Quantity
  • Availability
  • Add to Cart