ENA300792272 Enamine

2-(methylamino)-2-phenylacetic acid hydrochloride

  • MDL number MFCD06795795
  • Cas Number : 28544-42-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 242...244
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67524

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67524
Availability details for
Quantity
  • Availability
  • Add to Cart