ENA308583163 Enamine

N-(1-benzyl-1H-pyrazol-5-yl)-2-chloroacetamide

  • MDL number MFCD17167087
  • Cas Number : 1258641-39-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24681

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24681
Availability details for
Quantity
  • Availability
  • Add to Cart