ENA308583168 Enamine

2-chloro-N-{[4-(propan-2-yloxy)phenyl]methyl}acetamide

  • MDL number MFCD17167091
  • Cas Number : 81494-09-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-47368

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-47368
Availability details for
Quantity
  • Availability
  • Add to Cart