ENA308583169 Enamine

2-chloro-N-[(3,5-difluorophenyl)methyl]acetamide

  • MDL number MFCD17167092
  • Cas Number : 1258641-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-47507

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-47507
Availability details for
Quantity
  • Availability
  • Add to Cart