ENA308583172 Enamine

2-chloro-N-[(3-phenoxyphenyl)methyl]acetamide

  • MDL number MFCD17167095
  • Cas Number : 372137-82-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-47957

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-47957
Availability details for
Quantity
  • Availability
  • Add to Cart