ENA308583180 Enamine

1-{1-[4-(propan-2-yl)phenyl]ethyl}hydrazine hydrochloride

  • MDL number MFCD17167102
  • Cas Number : 1258650-52-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49497

Safety Information

msds https://www.enaminestore.com/catalog/EN300-49497
Availability details for
Quantity
  • Availability
  • Add to Cart