ENA308583181 Enamine

5-methyl-1H-pyrazole-4-sulfonyl chloride

  • MDL number MFCD17167103
  • Cas Number : 1179918-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-50519

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50519
Availability details for
Quantity
  • Availability
  • Add to Cart