ENA308583188 Enamine

1-[4-(1-aminoethyl)phenyl]pyrrolidin-2-one

  • MDL number MFCD17167108
  • Cas Number : 1258641-00-7
  • Molecular Weight : 204.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-56859

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56859
Availability details for
Quantity
  • Availability
  • Add to Cart