ENA308583195 Enamine

5-[(tert-butoxy)carbonyl]-1-phenyl-4,5-dihydro-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD17480351
  • Cas Number : 1258641-25-6
  • Molecular Weight : 290.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-63218

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63218
Availability details for
Quantity
  • Availability
  • Add to Cart