ENA308583196 Enamine

5-methyl-N-(pyrrolidin-3-ylmethyl)pyridin-2-amine

  • MDL number MFCD17167114
  • Cas Number : 1258650-60-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63230

Safety Information

msds https://www.enaminestore.com/catalog/EN300-63230
Availability details for
Quantity
  • Availability
  • Add to Cart