ENA308583199 Enamine

1-methyl-5-(morpholin-4-yl)-3-(propan-2-yl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD17480353
  • Cas Number : 1258650-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-63790

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63790
Availability details for
Quantity
  • Availability
  • Add to Cart