ENA308583210 Enamine

5,7-dimethyl-1H,2H,4H,5H,6H,7H-pyrazolo[1,5-a]pyrimidin-2-one

  • MDL number MFCD17167125
  • Cas Number : 1258640-91-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66358

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66358
Availability details for
Quantity
  • Availability
  • Add to Cart