ENA308583216 Enamine

N-[(5-methylfuran-2-yl)methyl]-2H-indazol-6-amine

  • MDL number MFCD17167131
  • Cas Number : 1157654-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66722
Availability details for
Quantity
  • Availability
  • Add to Cart