ENA308583217 Enamine

1,6-dimethyl-4-(trifluoromethyl)-1H,2H,3H,4H,5H,6H,7H-pyrazolo[3,4-b]pyridin-3-one

  • MDL number MFCD17167132
  • Cas Number : 1258641-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66812

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66812
Availability details for
Quantity
  • Availability
  • Add to Cart