ENA308583223 Enamine

1-(4-chlorophenyl)-3-phenoxypropan-1-amine hydrochloride

  • MDL number MFCD17167138
  • Cas Number : 1258641-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67283

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67283
Availability details for
Quantity
  • Availability
  • Add to Cart