ENA308583225 Enamine

6-amino-6-phenylhexanoic acid hydrochloride

  • MDL number MFCD17480356
  • Cas Number : 1258641-21-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145...147
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67345

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67345
Availability details for
Quantity
  • Availability
  • Add to Cart