ENA308583230 Enamine

(4-{2-[(4-chlorophenyl)sulfanyl]ethoxy}phenyl)methanamine hydrochloride

  • MDL number MFCD17167144
  • Cas Number : 1258641-46-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67641

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67641
Availability details for
Quantity
  • Availability
  • Add to Cart