ENA308583231 Enamine

2-(2-chlorophenyl)-2-(methylamino)acetic acid hydrochloride

  • MDL number MFCD17167145
  • Cas Number : 1258639-29-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-67672

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67672
Availability details for
Quantity
  • Availability
  • Add to Cart