ENA308583233 Enamine

3-hydrazinyl-8-methyl-8-azabicyclo[3.2.1]octane dihydrochloride

  • MDL number MFCD19442668
  • Cas Number : 110406-91-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68202

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68202
Availability details for
Quantity
  • Availability
  • Add to Cart