ENA308583234 Enamine

2-(chloromethyl)-4-fluoro-1H-1,3-benzodiazole hydrochloride

  • MDL number MFCD17167149
  • Cas Number : 1258649-53-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68307

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68307
Availability details for
Quantity
  • Availability
  • Add to Cart