ENA308583238 Enamine

2-{[2-(thiophen-2-yl)ethyl]amino}acetic acid hydrochloride

  • MDL number MFCD17167152
  • Cas Number : 1258640-11-7
  • Molecular Weight : 221.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68318

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68318
Availability details for
Quantity
  • Availability
  • Add to Cart