ENA308583239 Enamine

1-(5-chloropentyl)-1H-1,2,4-triazole hydrochloride

  • MDL number MFCD17167154
  • Cas Number : 1258650-18-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form oil
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68323

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68323
Availability details for
Quantity
  • Availability
  • Add to Cart