ENA308583240 Enamine

1-(5-chloropentyl)-3,5-dimethyl-1H-1,2,4-triazole hydrochloride

  • MDL number MFCD17167155
  • Cas Number : 1258649-57-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form oil
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68327

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68327
Availability details for
Quantity
  • Availability
  • Add to Cart