ENA308583246 Enamine

N-[(2,3-dihydro-1H-inden-5-yl)methyl]cyclopentanamine hydrochloride

  • MDL number MFCD17167161
  • Cas Number : 1258641-08-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68372

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68372
Availability details for
Quantity
  • Availability
  • Add to Cart