ENA308583247 Enamine

3-[(2-aminophenyl)amino]propanamide hydrochloride

  • MDL number MFCD17167163
  • Cas Number : 1258650-84-8
  • Molecular Weight : 215.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-68389

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68389
Availability details for
Quantity
  • Availability
  • Add to Cart