ENA308583253 Enamine

7-fluoro-2,3,4,5-tetrahydro-1,4-benzothiazepine hydrochloride

  • MDL number MFCD17167168
  • Cas Number : 1258650-31-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 269...271
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68425

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68425
Availability details for
Quantity
  • Availability
  • Add to Cart