ENA308583255 Enamine

3-(methoxycarbonyl)-1-phenyl-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD17167170
  • Cas Number : 1258650-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 171-172
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68437

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68437
Availability details for
Quantity
  • Availability
  • Add to Cart