ENA308583256 Enamine

2-(piperazin-1-ylmethyl)quinazolin-4-amine trihydrochloride

  • MDL number MFCD17167172
  • Cas Number : 1258639-47-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 276...278
Purity 95
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68467

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68467
Availability details for
Quantity
  • Availability
  • Add to Cart