ENA308583260 Enamine

2-[(prop-2-en-1-yl)carbamoyl]-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD17167176
  • Cas Number : 1258650-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-68496

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68496
Availability details for
Quantity
  • Availability
  • Add to Cart