ENA308583263 Enamine

2-(thiophen-2-yl)pent-4-yn-2-ol

  • MDL number MFCD17167179
  • Cas Number : 90674-24-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-68506

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68506
Availability details for
Quantity
  • Availability
  • Add to Cart