ENA308583266 Enamine

2-[(5-amino-3-bromo-1-methyl-1H-pyrazol-4-yl)formamido]acetic acid

  • MDL number MFCD17167183
  • Cas Number : 1258650-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68568

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68568
Availability details for
Quantity
  • Availability
  • Add to Cart