ENA308583268 Enamine

2-{4H,6H,7H-thieno[3,2-c]pyran-4-yl}acetic acid

  • MDL number MFCD17167185
  • Cas Number : 1258639-48-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68577

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68577
Availability details for
Quantity
  • Availability
  • Add to Cart