ENA308583272 Enamine

2-{5,7-dimethyl-2-oxo-1H,2H-pyrazolo[1,5-a]pyrimidin-6-yl}acetic acid

  • MDL number MFCD17480360
  • Cas Number : 1258639-52-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68649

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68649
Availability details for
Quantity
  • Availability
  • Add to Cart