ENA308583273 Enamine

1-(2-aminoacetyl)-N-methylpyrrolidine-2-carboxamide hydrochloride

  • MDL number MFCD17167189

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68650

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68650
Availability details for
Quantity
  • Availability
  • Add to Cart