ENA308583275 Enamine

1-(furan-3-carbonyl)piperazine hydrochloride

  • MDL number MFCD17167191
  • Cas Number : 1258649-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-68662

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68662
Availability details for
Quantity
  • Availability
  • Add to Cart