ENA308583276 Enamine

1-(5-chloropentyl)-4-methyl-1H-pyrazole hydrochloride

  • MDL number MFCD17167192
  • Cas Number : 1258649-84-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68680

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68680
Availability details for
Quantity
  • Availability
  • Add to Cart