ENA308583279 Enamine

2-chloro-1-{4-[(4-methoxyphenyl)methyl]piperazin-1-yl}ethan-1-one hydrochloride

  • MDL number MFCD17167195
  • Cas Number : 1258640-48-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68698

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68698
Availability details for
Quantity
  • Availability
  • Add to Cart