ENA308583283 Enamine

3-(chloromethyl)-1,2-dihydroisoquinolin-1-one

  • MDL number MFCD17167199
  • Cas Number : 1258640-51-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-68730

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68730
Availability details for
Quantity
  • Availability
  • Add to Cart