ENA308583284 Enamine

2-(ethylamino)-3-methylbutanoic acid hydrochloride

  • MDL number MFCD17167200
  • Cas Number : 1396964-36-5
  • Molecular Weight : 181.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68736

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68736
Availability details for
Quantity
  • Availability
  • Add to Cart