ENA308583287 Enamine

4-(4-methyl-2,5-dioxoimidazolidin-4-yl)benzamide

  • MDL number MFCD17480361
  • Cas Number : 1258650-55-3
  • Molecular Weight : 233.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-68774

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68774
Availability details for
Quantity
  • Availability
  • Add to Cart