ENA308583288 Enamine

2-chloro-1-{4-[(4-methylphenyl)methyl]-1,4-diazepan-1-yl}ethan-1-one hydrochloride

  • MDL number MFCD17167203
  • Cas Number : 1258649-61-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68775

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68775
Availability details for
Quantity
  • Availability
  • Add to Cart