ENA308583291 Enamine

3-(aminomethyl)-N-(butan-2-yl)benzamide hydrochloride

  • MDL number MFCD17167206
  • Cas Number : 1258650-62-2
  • Molecular Weight : 242.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-68801

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68801
Availability details for
Quantity
  • Availability
  • Add to Cart